Learn More
Description
Includes
Latex reagent, disposable slides (six-well), and prediluted positive and negative controls
- CLIA Complexity: Non-Waived
- CPT Code: 86308
- Latex agglutination slide test designed for the qualitative and semi-quantitative detection of infectious mononucleosis heterophile antibodies in serum and plasma
- Test results in three minutes
- Sensitivity of 99% and specificity of 99.4%
- For in vitro diagnostic use
Specifications
Specifications
| Clia Complexity | Non-waived |
| Detectable Analytes | Mononucleosis Heterophil Antibodies |
| Type | Mononucleosis Latex Test Kit |
| Includes | Latex reagent, disposable slides (6-well) and prediluted positive and negative control |
| Quantity | 150 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.