Description
Includes
Test kits include latex reagent, disposable slides (six-well), prediluted positive and negative controls, and package insert
- CLIA Complexity: Non-Waived
- CPT Codes: 86430/86318
- Latex agglutination test for the qualitative and semi-quantitative determination of rheumatoid factor in serum
- Can be used to both screen for levels of RF in serum greater than 10 IU/mL and determine titers of RF in serum in IU/mL using the semi-quantitative technique
- For in vitro diagnostic use
Specifications
Specifications
| CPT Code | 86430, 86318 |
| Clia Complexity | Moderate |
| Detectable Analytes | Rheumatoid Factor (RF) |
| Type | RF Rapid Latex Test Kit |
| Includes | Latex reagent, disposable slides (six-well), prediluted positive and negative control |
| Quantity | 50 Tests |
| Test Time | 2 min. |