Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Biogas Kit
SCI_ED

Catalog No. s02642 Shop All Fisherbrand Products
Change view
Click to view available options
Biogas kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S02642 Biogas kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Choking hazard (1) Small parts. Not for children under 3 yrs.

Catalog No. S02642
Add to Cart
Add to Cart

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Biogas digester
  • 1 gal. biogas collection bag
  • All necessary fittings and tubing
  • Check valve
  • Combustion nozzle assembly
  • Personal safety gear for one person (goggles, gloves, apron)

Requires

Approximately 1/2 gallon of antibiotic-free manure as your biomass

Create real bio-renewable energy using readily available organic waste materials.

  • Biogas can be used to power engines, generate electricity, cook food and heat homes
  • Simply vary the biomass source and/or digestion temperature, and you have a variety of comparative experiments to analyze
  • Significant gas production in about three to five days
  • Rabbit manure works well
  • Grades 8-14
  • Class size: 24 students

Specifications

Product Type Biogas Kit
Includes Biogas digester, 1 gal. biogas collection bag, check valve, combustion nozzle assembly, personal safety gear for one person (goggles, gloves, apron), all necessary fittings and tubing
For Use With (Application) Create real bio-renewable energy using readily available organic waste materials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.