Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuSkan™ FC Filter-Based Microplate Photometer

Catalog No. 14377575 Shop All Fisherbrand Products
Change view
Click to view available options
Description:
accuSkan FC
accuSkan FC with incubator
Plate Type:
96 and 384-well Plates
96-well Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Plate Type
14-377-575 accuSkan FC 96-well Plate
14-377-576 accuSkan FC with incubator 96 and 384-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-377-575

For research and routine ELISA applications

A proven ELISA solution—accuSkan FC and the Fisher Scientific accuWash washer. The combination of the accuSkan FC microplate photometer and the accu-Wash™ microplate washer offers a convenient modular solution for routine and research ELISA applications.

A proven ELISA solution—accuSkan FC and the Fisher Scientific accuWash washer. The combination of the accuSkan FC microplate photometer and the accu-Wash™ microplate washer offers a convenient modular solution for routine and research ELISA applications.
  • Broad wavelength range of 340 to 850nm for a wide variety of applications
  • Eight-position filter wheel with three standard pre-installed filters (405, 450 and 620nm)
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • On-board linear shaking and optional incubation up to 50°C for temperature critical assays
  • Superior usability with both internal software and dedicated PC software
  • Data export capability via USB memory stick for easy data transfer
Applications: Immunoassays (ELISA), protein and enzyme assays,cytotoxicity and proliferation assays, growth curves and endotoxins

Warranty and Services

1 Year

TRUSTED_SUSTAINABILITY

Specifications

Detector Type Photodetector
Dimensions (L x W x H) 11.4 x 15.7 x 8.7 in. (29 x 40 x 22 cm)
Interface PC Control, Internal Software
Light Source Quartz Halogen Lamp
Measurement Speed 96 wells: 6 sec. (fast mode), 12 sec. (normal mode)
Plate Type 96-well Plate
Shaking Linear shaking with 3 modes
Accuracy ±1% (0 to 3 Abs) or ±0.003 Abs, which ever is greater at 405 nm
Description accuSkan FC
Electrical Requirements 100/240 V, 50/60 Hz
Excitation Filters 8-position filter wheel; filters installed: 405nm, 450nm, and 620nm
For Use With (Equipment) Robotic automation
Linearity 96-well plate: 0 to 3 Abs (±2% at 405 nm)
384-well plate: 0 to 2.5 Abs (±2% at 405 nm)
Precision CV ≤ 0.2% (0.3 to 3 Abs) at 405 nm
Resolution 0.001 Abs
Warranty 1 Year
Wavelength Range 340 to 850 nm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.