Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cheesecloth Wipes
SCI_ED

Catalog No. S04824 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
18 x 36 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S04824 18 x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S04824

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Reagent-grade quality Cheesecloth Wipes.

  • 100% brilliant white, pure cotton construction will not scratch sensitive surfaces Have a high degree of cleanliness, softness, absorbency and strength
  • Can be used to safely clean microscope and camera optical parts, as well as delicate glassware, syringes, pipets, and pH electrodes
  • Absorb six times their weight in liquids and are useful for mopping up solvents and aqueous solutions
  • Inert to even harsh solvents; good filtering material
  • Grade 10 (mesh size)

Specifications

Product Type Cheesecloth Wipe
Length (English) 18 in.
Length (Metric) 457.2 mm
Width (English) 36 in.
Width (Metric) 914.4 mm
Material Cotton
Dimensions (L x W) 18 x 36 in.
For Use With (Application) For used to clean microscopes, optical surfaces, glassware, pH electrodes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.