Learn More
Fisher Scientific™ ChemExpress™ Electronic MSDS System
MSDS information at your fingertips!
Description
CD-ROM provides critial safety information on over 3000 analytical laboratory reagents. All MSDSs are in the internationally recognized 16-section ANSI standard format, so finding MSDSs is quick and easy. An updated CD-ROM is sent every three months.
- Features searches by full or partial chemical name, catalog number, molecular formula, and CAS number of pure product or component
- System includes six additional languages (French, Spanish, German, Italian, Dutch, and Swedish), and also allows additional MSDSs to be added
- Compatible with all versions of Windows™ (Minimum system requirements for operation: Pentium™ processor, 32MB RAM, CD-ROM drive and mouse, hard drive with 100-600MB free space, and monitor with VGA or higher resolution)
- May be installed to hard drive for faster performance, or run from the CD
- Fisher/Acros™ Organics/Maybridge Version includes same features as Fisher Chemical edition, but also provides access to Acros Organics and Maybridge MSDSs for 26,000 organic and analytical laboratory reagents
- Both editions available configured for single user, or for network that can be accessed by an unlimited number of users
Specifications
CD-ROM | |
Electronic MSDS System | |
Provides access to MSDS for Fisher Chemicals |