Learn More
Fisherbrand™ Computer Data Acquisition System for Digital Barometer
Catalog No. 14650120 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand Traceable Digital Barometer
Catalog No. For Use With (Equipment)
14-650-120 Fisherbrand Traceable Digital Barometer
1 options
Catalog No. 14-650-120
Add to Cart
Add to Cart
Keep track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more. Computer Data Acquisition System for Digital Barometer is for use with Fisherbrand™ Traceable™ Digital Barometer.
  • Allows up to eight unique Traceable™ meters to simultaneously capture data and send it to directly to email to any smartphone or computer
  • Permanently capture readings; keep track of results
  • Readings are saved to a file that can be viewed, printed as-is, or merged in a report or spreadsheet format
  • Data continues to be collected in background even when user works on other software programs
  • 24/7 audible/visual alarm reminds user to check on data
Specifications
Type Data Acquisition Software
Display Type No Display
Software Type Computer Data Acquisition System
Includes CD, 6 ft. Cable
For Use With (Application) Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more
For Use With (Equipment) Fisherbrand Traceable Digital Barometer
Length (English) Cable 6 ft.