Learn More
Fisherbrand™ Electrophotometer II Colorimeter Square Micro Absorption Cell
Shop All Fisherbrand Products
Description
- Optically flat
- Parallel
- Two sides clear
- Two sides frosted
Specifications
Specifications
| Type | Microabsorption Cell |
| For Use With (Equipment) | Electrophotometer II colorimeter |
| Pathlength | 9.97 mm |
| Shape | Square |