Learn More
Fisherbrand™ ER1425S-HT™ High-Temperature Datalogger Battery
SDP
Catalog No. 15060079 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
HiTemp140 and PR140 Dataloggers
Catalog No. For Use With (Equipment)
15-060-079 HiTemp140 and PR140 Dataloggers
1 options
Catalog No. 15-060-079
Add to Cart
Add to Cart
3.6V lithium battery
For use with Fisherbrand™ HiTemp140 (Cat. No. 15-060-080) and Fisher Scientific™ High-Temperature Pressure Datalogger (Cat. No. 15-060-100).
  • 150°C, 3.6V lithium battery
  • 1/2 AA pressure contact
Specifications
Type Battery, Datalogger
For Use With (Equipment) HiTemp140 and PR140 Dataloggers
Temperature Range (English) 302°F
Temperature Range (Metric) 150°C