Learn More
Fisherbrand™ High Purity Hydrogen Gas Generators For GC Fuel Gas Accessory, PM Kit
SDP
Catalog No. p-6771198 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
24 Month PM Kit
6 Month PM Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-950-644 6 Month PM Kit
14-950-645 24 Month PM Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-950-644
Only null left
Add to Cart
Add to Cart
Includes
  • Six Month Kit: DI Cartridge, In-line Water filter, and 3 Environmental Tank Vent Fliters
  • 24 Month Kit: Desiccant Cartridge, DI Cartridge, In-line Water filter, 3 Environmental Tank Vent Fliters, Water Pump and Service Dongle
For use with High Purity Hydrogen Gas Generators for GC Fuel Gas.
Six month PM Kit
  • Helps ensure quality of deionized water in Fisher Scientifc™ HP range of H2 generators maximizing life of PEM cell
24 month PM Kit
  • Required at 16,000 hours of operation when Fisher Scientific HP generator will alarm indicating that service is due
Specifications
Type 6 Month PM Kit
Description Six Momth PM Kit for Fisher Scientific High Purity H2 Generator
Includes 1 DI cartridge, 1 in-line water filter and 3 environmental filters
For Use With (Equipment) High Purity Hydrogen Gas Generators For GC Fuels
For Use With (Application) Gas chromatography, fuel gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), Electrolytic Conductivity Detector (ELCD & Hall ELCD), other combustion detectors