Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Scientific™ Bath Circulator Accessories: Plumbing Packages

Plumbing kits for use with bath circulators and chillers

$589.00 - $660.00

Specifications

Temperature Range -25°C to +100°C
View More Specs

Products 2
Catalog Number Type Price Quantity & Availability  
Catalog Number Type Price Quantity & Availability  
13-265-215
View Documents
Plumbing Kit, Insulated
Each for $660.00
Only null left
Add to Cart
 
13-265-214
View Documents
Plumbing Kit, Uninsulated
Each for $589.00
Only null left
Add to Cart
 
Description

Description

Plumbing kits for use with bath circulators and chillers

  • Plumbing kit
  • Material: Viton
Specifications

Specifications

-25°C to +100°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.