Learn More
Thermo Scientific™ Thermo Scientifi™ Chiller External Pressure Reducers
External pressure reducers
$709.50 - $741.00
Specifications
Type External Pressure Reducer (EPR)
Size 1/2 in.
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
13-265-234
View Documents
Thermo Scientific™
196000000001
ThermoChill III
Each for $741.00
Only null left
Add to Cart
13-265-233
View Documents
Thermo Scientific™
196000000000
ThermoChill I and II
Each for $709.50
Only null left
Add to Cart
Description
Description
External pressure reducers for ThermoChill Recirculating chillers
  • 1/2 in. external pressure reducer
  • Adjustable to safely reduce or limit system pressure and flow
Specifications
Specifications
External Pressure Reducer (EPR)
1/2 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.