Learn More
Fisherbrand™ Isotemp™ Refrigerated/Heated Bath Circulators: 5.4-6.5L, 115V/60Hz
GSA_VA
For laboratory and industrial applications.
 View more versions of this product
Catalog No. 13-874-674
Only null left
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Description
Description
Heated Bath Circulators: (Ambient +10°C to +200°C) A complete system made up of an immersion circulator and a bath. Use as a temperature controlled bath or to circulate fluid externally to an application.
Isotemp heated bath circulators feature reliable, high-wattage heaters that deliver rapid fluid heat up for your application. Standard high temperature cutout (HTC) circuitry ensures application temperatures do not exceed user selectable limits.
Low-profile model enables easy bath access when lab counter height presents a challenge. Seamless stainless-steel tanks guarantee durability and easy cleaning. Robust refrigeration system ensures that samples and applications can be cooled quickly. Fine-tuned refrigeration metering allows for excellent temperature stability at all times.
  • All bath drains are located on the front of the unit with an integrated valve
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.79 to 1.58 gal.
-20°C to +100°C
External circulation plumbing with 8 and 12mm fitting, supply and return clamps, work area cover
6200 R20
±0.05°C
16.4 in.
8 in.
25.2 in.
60 lb.
115 V 60 Hz
60 Hz
3 to 6 L
1.2 Kw
805 mbar
21 gpm
41.6 x 20.3 x 64 cm
41.6 cm
20.3 cm
64 cm
27.2 kg
115 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.