Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Long-Stem Digital Thermometer
SCI_ED

Catalog No. s407993 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S407993 -50°C to +150°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S407993
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Durable and safe, containing no mercury or alcohol. Fisherbrand Long-Stem Digital Thermometers are rugged and versatile digital thermometers to read soil, or stir and measure the temperature of solutions. Can also be used with incubators.

  • Accuracy: ±1°C
  • Large, bright 0.2 in. (6mm) display
  • Reads melting/freezing point temperatures, heat of fusion, temperatures hotter than 110°C (230°F)
  • Stainless-steel probe is resistant to acids, bases and solvents
  • Dual temperature range: -50° to +150°C (-58° to +302°F)
  • Stem length: 8 in. (20.3cm); diameter: 3mm
  • Overall length: 11 in. (28cm)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.14 in.
For Use With (Application) Food, gases, samples, reagents, water baths and semi solids
Includes Stainless Steel probe traceable certificate, protective sleeve, battery
Length (English) Probe 8 in.
Length (Metric) 280 mm
Length (Metric) Probe 20 cm
Material ABS Plastic
Resolution 0.1°C
Product Type Digital Thermometer
Length (English) 11 in.
Max. Temperature (English) 302°F
Max. Temperature (Metric) 150°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +150°C
Calibrated Yes
Accuracy ±1°C
Display Digital LCD
Width (Metric) 8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.