Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Scientific™ Tissue Path™ Tissue Cassette/Ring Storage Cabinet/Box: Bench Stand and Insert Trays

Catalog No. 22025589
Change view
Click to view available options
Product Type:
Bench Stand
Insert Tray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-025589 Insert Tray
22-033256 Bench Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-025589 Supplier National Scientific™ Supplier No. HABET20

Includes

Trays only. Bench Stand is sold separately.

Insert Tray and Bench Stand for use with Fisher Scientific Tissue Path Tissue Cassette Embedding Ring Storage Cabinet and Fisher Scientific Tissue Path Tissue Cassette Storage Box

  • Bench Stand Dimensions (L x W): 13.125 x 9.375 in.
  • Height: 4.75 in. (back); 1.125 in. (front)

Specifications

Product Type Insert Tray
Color Transparent
For Use With (Equipment) Fisher Scientific Tissue Path
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.