Learn More
National Scientific™ Tissue Path™ Tissue Cassette/Ring Storage Cabinet/Box: Bench Stand and Insert Trays
Catalog No. 22033256
Change view
Click to view available options
Product Type:
Bench Stand
Insert Tray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-033256 Bench Stand
22-025589 Insert Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-033256 Supplier National Scientific™ Supplier No. HATBS01
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Includes
Bench Stand only. Trays are sold separately.
Insert Tray and Bench Stand for use with Fisher Scientific Tissue Path Tissue Cassette Embedding Ring Storage Cabinet and Fisher Scientific Tissue Path Tissue Cassette Storage Box
  • Bench Stand Dimensions (L x W): 13.125 x 9.375 in.
  • Height: 4.75 in. (back); 1.125 in. (front)
Specifications
Product Type Bench Stand
Holds Insert Trays
For Use With (Equipment) Fisher Scientific Tissue Path