Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Traceable™ One-Shot™ pH Buffer Standards pH Certified Reference Material (CRM), Fisherbrand™

Catalog No. 15078288 Shop All Fisherbrand Products
Change view
Click to view available options
pH:
10.012
4.005
4.005, 7.000, 10.012
7.00
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
15-078-288 4.005, 7.000, 10.012
15-078-287 10.012
15-078-285 4.005
15-078-286 7.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 15-078-288
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Performs accurate, reproducible pH calibrations with color-coded bottles. Traceable™ Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.

  • Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
  • 100% compatible with all makes of pH instruments and electrodes
  • Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
  • Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
  • Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
  • Supplied with individual temperature chart, traceability information and Traceable™ Certificate

Specifications

Chemical Name or Material One-Shot pH Buffer Standards (CRM) 6-Pack Assortment
Color Undesignated
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.005, 7.000, 10.012
Physical Form Liquid
Quantity 6 x 100 mL
Solution Type Buffer
Packaging 100 mL One-Shot (2 of each)
Traceability to NIST Certified and Traceable™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.