Learn More
Fisherbrand™ Ultra High Purity Hydrogen Gas Generators Accessory, Preventative Maintenance Kit
SDP
Catalog No. 14950652 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
24 Month PM Kit
6 Month PM Kit
60 Month PM Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14-950-652 60 Month PM Kit
14-950-650 6 Month PM Kit
14-950-651 24 Month PM Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-950-652
Add to Cart
Add to Cart
Includes
  • 60 Month Kit: Replacement Dryer Columns and Service Dongle
For use with Ultra High Purity Hydrogen Gas Generators for GC Carrier and Fuel Gas.
60 month PM Kit
  • Required at 40,000 hours of operation when Fisher Scientific UHP generator will alarm indicating that service is due
Specifications
Product Type 60 Month PM Kit
Description 60 Month PM Kit for Fisher Scientific Ultra High Purity H2 Generator
Includes Dryer column assemblies (X2)
For Use With (Application) Gas chromatography, fuel gas, carrier gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), nitrogen phosphorous detector carrier gas (NPD), thermal conductivity detector carrier gas (TCD), Electrolytic Conductivity Detector reactio
For Use With (Equipment) Ultra High Purity Hydrogen Gas Generators For GC Carrier and Fuel Gas