Learn More
Fisher Scientific™ Zero Air Generators For GC Fuel Accessory, Annual PM Kit
SDP
Catalog No. 14950664 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Annual PM Kit
Catalog No. Product Type
14-950-664 Annual PM Kit
1 options
Catalog No. 14-950-664
For use with Zero Air Generators For GC Fuel
  • Helps protect catalyst module and downstream applications form contamination
  • One year PM filter Kit
Specifications
Product Type Annual PM Kit
For Use With (Application) Gas chromatography, flame support gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD)
For Use With (Equipment) Zero Air Generators For GC Fuel (14950556 and 14950663)
Includes Inlet and Outlet filters