Learn More
Fisherbrand™ DNA Sequencing Systems: Combs for Model FB-SEQ-3545
Catalog No. FBSEQ604 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Owl™ S3S Aluminum-Backed Sequencer System
Catalog No. For Use With (Equipment)
FB-SEQ-604 Owl™ S3S Aluminum-Backed Sequencer System
1 options
Catalog No. FB-SEQ-604
Eliminate comb assembly with these heavy-duty, one-piece combs.
  • For use with model FB-SEQ-3545
  • Heavy duty, one piece combs eliminate comb assembly
TRUSTED_SUSTAINABILITY
Specifications
Description Well Comb
For Use With (Equipment) Owl™ S3S Aluminum-Backed Sequencer System
Product Type Combs
Teeth Count 60
Thickness (Metric) Teeth 0.4 mm
Volume (Metric) Well 9μL