Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System: Alumina Plate

Catalog No. FBVE1015 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
FisherBiotech FB-VE10-1 Vertical Electrophoresis System; Owl P7-CST Multiple Gradient Gel Caster
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
FB-VE10-15 FisherBiotech FB-VE10-1 Vertical Electrophoresis System; Owl P7-CST Multiple Gradient Gel Caster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB-VE10-15

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

For use with FB-VE 10-1 vertical electrophoresis systems

Notched alumina; 10L x 10cmW.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) FisherBiotech FB-VE10-1 Vertical Electrophoresis System; Owl P7-CST Multiple Gradient Gel Caster
Type Notched Alumina Plate
Material Alumina
Length (Metric) 10 cm
Width (Metric) 10 cm
Height (English) 0.39 in.
Height (Metric) 1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.