Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Large Gel Electrophoresis System
SCI_ED

Catalog No. S98616 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
14.7 x 11 x 4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S98616 14.7 x 11 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98616

Includes

Three-point leveling platform, buffer chamber, safety lid with attached power supply leads, UV-transparent gel tray, nonskid rubber feet and three 1.5mm combs (16-, 24-, 36-well)

Provide leak-free casting in a 20 x 25cm gel tray with six comb positions.

Improve resolution in Northern and Southern blotting of large gels and multiple samples.

  • Available with same self-contained buffer recirculation method as Midigel System (S98617)
  • Run up to 36 samples per comb position with supplied combs
  • Permit up to four sets of samples to be run equal distances on the same gel
  • Allow shorter gels to be poured when used with accessory wall comb
  • Buffer volume: 2.3L
  • May also be used with programmable power inverter for field inversion electrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Type Standard Gel Electrophoresis System
Includes Three point leveling platform, Buffer chamber, safety lid with attached power supply Leads, UV transparent gel tray, nonskid Rubber feet and three 1.5mm comb
Model Large gel system
Volume (English) Buffer 77.7 oz.
Volume (Metric) Buffer 2300 mL
Dimensions (L x W x H) 14.7 x 11 x 4 in.
Length (English) 14.7 in.
Length (English) Gel 9.8 in.
Length (Metric) 37.5 cm
Length (Metric) Gel 25 cm
Width (English) 11 in.
Width (English) Gel 7.8 in.
Width (Metric) 28 cm
Width (Metric) Gel 20 cm
Height (English) 4 in.
Height (Metric) 10 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.