Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Midigel Electrophoresis System
SCI_ED

Catalog No. s98617 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
9.6 x 7 x 3.12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S98617 9.6 x 7 x 3.12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S98617
This item is not returnable. View return policy

Includes

Buffer chamber, safety lid with attached power supply leads, UV-transparent gel tray, nonskid rubber feet and two 1.5mm thick combs (12- and 20-well)

Provide leak-free gel casting in 13 x 16cm gel tray with two comb positions.

Run up to 32 samples on one gel with combs supplied.

  • Available with buffer recirculation method that prevents ionic depletion on long gel runs, keeps ionic gradients from distorting results when multiple sample series are run simultaneously and prevents pH changes that can cause problems in RNA gels
  • Buffer volume: 800mL
  • Can run up to 48 samples on a gel with accessory combs

TRUSTED_SUSTAINABILITY

Specifications

Type Standard Midigel Electrophoresis System
Includes Buffer chamber, safety lid with attached power supply Leads, UV transparent gel tray, nonskid Rubber feet and two 1.5mm thick combs (12 and 20-well)
Model Midigel system
Volume (English) Buffer 27 oz.
Volume (Metric) Buffer 800 mL
Dimensions (L x W x H) 9.6 x 7 x 3.12 in.
Length (English) 9.6 in.
Length (English) Gel 6.2 in.
Length (Metric) 24.5 cm
Length (Metric) Gel 16 cm
Width (English) 7 in.
Width (English) Gel 5.1 in.
Width (Metric) 18 cm
Width (Metric) Gel 13 cm
Height (English) 3.12 in.
Height (Metric) 8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.