Learn More
Fisherbrand™ Vertical Electrophoresis Multiple Gradient Gel Caster
Catalog No. FBMGC1614 Shop All Fisherbrand Products
Catalog No. FBMGC1614
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Casts up to five gels simultaneously
  • Gradient gels can be poured by pumping acrylamide through a port on caster's base
  • Linear gels can be poured from the top of the unit with the port closed
  • Gels suited for use in FisherBiotech vertical electrophoresis systems or similar units
  • Silicone gasket provides leakproof seal
  • Casters disassemble easily for cleaning
TRUSTED_SUSTAINABILITY
Specifications
Type Caster