Learn More
Fisherbrand™ Wall Comb for Horizontal Midigel Systems
Catalog No. FBSB1337 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Owl A1 Large Gel Electrophoresis System
Catalog No. For Use With (Equipment)
FB-SB-1337 Owl A1 Large Gel Electrophoresis System
1 options
Catalog No. FB-SB-1337 Supplier Fisherbrand™ Supplier No. A1WALL
Wall Comb for Horizontal Midigel Systems
  • Wall comb permits casting of gel that is shorter than gel tray
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Type Wall Comb
Teeth Count 1
Width (English) Teeth 5.1 in.
Width (Metric) Teeth 13 cm