Learn More
Bio Plas 2mL Siliconized Screw Cap Tube
Description
Ideal for packing, transporting, and storing diagnostic tests, especially in the area of biotechnology, molecular biological products and reagents.
- Secure screw closure
- Ensures a safe transport with durable silicone
- Packages and stores samples
Specifications
Specifications
| Closure Material | Silicone |
| Capacity (Metric) | 2 mL |
| Color | Natural |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Type | Screw Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.