Learn More
Fisherbrand™ Animal Ear-Punch
Catalog No. 13812201 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
2 mm punch hole
Catalog No. Size
13-812-201 2 mm punch hole
1 options
Catalog No. 13-812-201
Add to Cart
Edge
Add to Cart
Fisherbrand Ear punch are used for punching holes into an animal's ear for labeling purposes.
Fisherbrand™ Animal Ear-Punch. These instruments are available in multiple designs, including thumb-style, plier-style, or scissors-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.
Ear-Punch
  • For use with animal ear tags
  • Thumb-type for punching holes into thin part of animal’s ear
  • Plier-type: Provides a longer handle for gripping and punching into animal ear
  • Made from corrosion-resistant ASTM (899-94) series 400 stainless steel
  • Available Ear-punch: Thumb, plier or scissor type
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
  • Electronically passivated for corrosion resistance
Specifications
For Use With (Application) Animal Identification
Type Ear Tag Punch
Size 2 mm punch hole