Description
Requires
13-812-201 punch to make hole in animal's ear (above right)
Ear-Punch
- For use with animal ear tags
- Thumb-type for punching holes into thin part of animal’s ear
- Plier-type: Provides a longer handle for gripping and punching into animal ear
- Made from corrosion-resistant ASTM (899-94) series 400 stainless steel
- Available Ear-punch: Thumb, plier or scissor type
- Compatible with Gamma/ETO/Autoclavable sterilization methods
- Electronically passivated for corrosion resistance
Ear-Tags
- Insert tag through hole; place washer behind hole
- Bend ends of tag back in rivet fashion, leaving space for ear growth
- Made from ASTM standard aluminum
Specifications
Specifications
| Type | Animal Tag |
| Includes | Washer |
| Material | Aluminum |