Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
SDP
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
 View more versions of this product
Catalog No. 11-603-3VC
Only null left
Add to Cart
Description
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
  • Use Cylinder 08-530K
  • Temperature of standardization: 60°F
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Compliance
Calibrated according to ASTM™ E 126 specifications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrometer
0.001
260 mm
0.800 to 0.850 ASTM 105H-62
60°F
105H
3-point Cert, ASTM E126, E100-14 T1
Liquids heavier than Water
260 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.