Learn More
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Description
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
- Use Cylinder 08-530K
- Temperature of standardization: 60°F
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Compliance
Calibrated according to ASTM™ E 126 specifications
Specifications
83H | |
3-point Cert, ASTM E126, E100-14 T1 | |
Liquids heavier than Water | |
275 mL |
0.0005 | |
330 mm | |
0.700 to 0.750 ASTM 83H-62 | |
60°F |