Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Caps for Multi-Purpose Sample Cups

Catalog No. 0254410 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-544-10 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-544-10

missing translation for 'validMainframeMsg'

Fit 0.25, 0.5 and 2.0mL capacity cups

  • Color: Natural

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Product Type Sample Cap
For Use With (Application) To prevent cracking or tipping
For Use With (Equipment) 0.25, 0.5 and 2 mL multipurpose sample cup
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.