Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ ColorFrost™ Microscope Slides with Clipped Corners

Catalog No. 22037162 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Green
Pink
Yellow
Corner Style:
45° Clipped
Clipped
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Corner Style
22-037-162 Pink 45° Clipped
22-037-160 Blue Clipped
22-037-161 Green Clipped
22-037-163 Yellow Clipped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-037-162
Add to Cart
Add to Cart
This item is not returnable. View return policy

Durable, opaque printed surface inert to common laboratory chemicals and reagents

  • 45° clipped corners; color-coded frosted end
  • Raised printed writing surface eliminates need for paper labels and tedious relabeling of slides
  • Autoclavable

Specifications

Length (Metric) 25 mm
Width (Metric) 75 mm
Thickness (Metric) 1 mm
Frosted Colorfrost
Color Pink
Coating Chemical Resistant
Corner Style 45° Clipped
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.