Learn More
Fisherbrand™ Applicators
Catalog No. 23400125 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Applicator
Length (English):
3 in.
6 in.
Sterility:
Non-sterile
Sterile
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (English) Sterility
23-400-125 Applicator 6 in. Sterile
23-400-119 Applicator 6 in. Non-sterile
23-400-124 Applicator 6 in. Sterile
23-400-101 Applicator 6 in. Non-sterile
23-400-100 Applicator 3 in. Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-400-125
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Purified cotton
  • Available sterile or nonsterile
  • With wooden or plastic handles
Applications:
  • Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instruments
Specifications
Quantity 1 per envelope
Length (Metric) 15 cm
Handle Material Wood
Tip Material Cotton
Sterility Sterile
Length (English) 6 in.
For Use With (Application) Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments
Product Type Applicator