Description
The restricted opening provides effective specimen closure and can help minimize exposure to droplets of blood or other potentially infectious materials during sample handling, preparation, processing, and storage.
- All-inclusive “closed” filtration system with a combination filter, transfer pipet, and cap
- Filters fibrin and gel particulates, dispenses serum or plasma, prevents evaporation, and helps protect ISE systems
- All-in-one system maintains sample integrity and eliminates transfer error
- Dispense minute or large amounts of sample without transfer pipets or add-on products
- Ideal for dispensing samples for instrumentation, pour-off or other use
- Use with glass collection tubes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Drops/mL | 50 drops/mL |
| For Use With (Equipment) | Use with Glass collection tubes |
| Quantity | 100 Pack |
| For Use With (Application) | Dispensing samples for instrumentation, pour-off or other use |
| Includes | Combination filter, transfer pipet, cap |
| Product Type | Dispense Filtration System |