Learn More
Fisherbrand™ Dispense + Serum Filter
Catalog No. 22267383 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
22-267-383 100 Pack
1 options
This item is not returnable. View return policy
Catalog No. 22-267-383
Add to Cart
Add to Cart
This item is not returnable. View return policy
Eliminates the need for disposable pipets, add-on products and capping materials.
  • One-step, self-contained filtration, dispensing and storage device used to deliver centrifuged serum or plasma
  • Restricted opening acts as a cap closure
  • Use with plastic collection tubes
Specifications
Quantity 100 Pack
Product Type Dispense Serum Filter
For Use With (Application) Plastic evacuated collection tubes (BD, Greiner, or Sarstedt), or 12 x 75mm or 13 x 100mm Polypropylene and Polystyrene Plastic test tubes
For Use With (Equipment) 12 x 75mm or 13 x 100mm Polypropylene and Polystyrene test tubes