Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Graduated Transfer Pipettes
SCI_ED

Catalog No. S05014 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity: 5.8 mL; Nonsterile; 500/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05014 Capacity: 5.8 mL; Nonsterile; 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05014

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for hematology, urinalysis, chemistry and blood banking — anywhere there is a need for quick, safe transfer of fluids.

Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from unbreakable, see-through low-density polyethylene to eliminate the hazards of broken glass and exposure to infectious materials.

  • Available gamma radiation-sterilized or nonsterile
  • 5.8mL capacity with 0.25 mL graduations
  • Draw 3.4 mL per squeeze
  • Inert to biological fluids and most acids
  • One squeeze draws from 2.0 to 3.3 mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)

Order Info

Available individually wrapped in peel-apart sleeve, or bulk packaged in easy-open bags of 10 or 20.

TRUSTED_SUSTAINABILITY

Specifications

Bulb Draw (Metric) 3.4 mL
Material Polyethylene
Sterility Non-sterile
Graduated Yes
Volume (Metric) 5.8 mL
Length (Metric) 15.4 cm
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.