Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ DOT Shipping Labels
Labels for miscellaneous hazardous materials and infectious substance
Description
- For class 9 miscellaneous dangerous goods or class 6 toxic (poisonous) and infectious substances
- Supplied in packs of 25
Specifications
HM-215B | |
Paper | |
For Class 9 Miscellaneous Dangerous Goods | |
25/Pk. | |
4 in. |
Label | |
DOT Shipping Label | |
The durable adhesive will stick to most flat, clean surfaces, Adhesive Vinyl signs will work for short term outdoor applications and in moderate temperature | |
4 in. | |
10.16 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction