Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Drawer Locks

Catalog No. 19033566 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-033-566 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-033-566
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with SS and GZ series drawers only

Help protect your drawer's contents, a locking drawer will give you greater security. For use with SS and GZ drawers only.

  • Key included
  • Color: Silver
  • Material: Stainless steel
  • Model: TA-14

Specifications

For Use With (Application) Securing drawers
Color Silver
No. of Doors 1
Includes Lock and key
Material Stainless Steel
Model TA-14
For Use With (Equipment) SS and GZ series drawer
Product Type Drawer Lock
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.