Learn More
Fisherbrand™ Engraved-Stem Mercury Laboratory Thermometers
Catalog No. 150411B Shop All Fisherbrand Products
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste.
Catalog No. 15-041-1B
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste.
For general testing and laboratory procedures where an ASTM™ or similar thermometer is not required
  • Supplied in easy-to-read yellow-backed glass with Permetch™ brand permanent pigmentation of graduations
Certifications
Conform to NIST™ Monograph No. 150.