Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
SDP
Catalog No. 1007557 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Application):
1000mL erlenmeyer flask
250mL Erlenmeyer flask
500 mL Erlenmeyer Flask
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
1007557 500 mL Erlenmeyer Flask
1007555 250mL Erlenmeyer flask
1007558 1000mL erlenmeyer flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1007557
Add to Cart
Add to Cart
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
  • Safe and secure attachment
  • Manufactured from strong and durable stainless steel
  • Provides superior stability
Specifications
Type Flask Clamp
For Use With (Equipment) Ultrasonic Cleaner
For Use With (Application) 500 mL Erlenmeyer Flask