Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ FE150 pH benchtop meter bio set with double junction gel pH electrode, ATC probe and stand
SCI_ED

Catalog No. s35924 Shop All Fisherbrand Products
Change view
Click to view available options
Range (pH):
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range (pH)
S35924 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S35924
Add to Cart
Add to Cart

Designed for convenient routine pH measurement in the lab: user-friendly, stores data sets in non-volatile memory and meter-attachable electrode stand. Efficient sample reading and recording with flexible display options: display both pH and temperature or mV and temperature simultaneously.

The essential functions needed for routine measurements
  • One to five point pH calibration
  • Automatic buffer recognition with selection of two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.3°C
Accuracy (pH) ±0.01
Includes FE150 meter, double junction gel pH electrode, ATC probe and stand
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Calibration Points 1
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Memory 100 data sets
Warranty 3 Years
Model FE150 pH Benchtop Meter Bio Set
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.