Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Flat and Domed 8-Cap Strips, Flat Cap Strips
GSA_VA

Catalog No. p-3599606 Shop All Fisherbrand Products
Change view
Click to view available options
Format:
Domed Cap Strips
Flat Cap Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14-230-230 Flat Cap Strips
14-230-231 Domed Cap Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-230-230
Only null left
Add to Cart
Add to Cart

Suitable for use with 96-well PCR plates and PCR tube strips

  • Eight caps per strip
  • Cleanroom produced
  • Virgin polypropylene
  • Flat caps offer large, labeled end tabs for easy removal and strip orientation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Color Natural
Format Flat Cap Strips
For Use With (Equipment) 96-well PCR Plates or PCR Tube Strips
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.