Learn More
Description
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
- Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
- Wide mouth 53 mm (2 in.) for easy refilling
- Color-coded polypropylene closures aid in quick and easy identification
- Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
- Bottle with Label: TOLUENE, Red LDPE bottle, does not include vented valve
- Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER, does not indicate hazardous chemical and does not have or require GHS pictogram or signal word
- Bottle with Label: METHYL ETHYL KETONE, does not include vented valve
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to Workplace Labels.
Specifications
Specifications
| Capacity | 500 mL |
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16.91 oz. |
| Color | Natural |
| Material | LDPE |
| Quantity | 3/Pk. |
| Sterility | Non-sterile |
| Closure Color | Yellow |
| Closure Included | Yes |
| Disposable | Reusable |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.