Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ GL45 Thread Tube Screw Cap
SDP

Catalog No. 13247GL45 Shop All Fisherbrand Products
Change view
Click to view available options
Closure Size:
45 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
13-247-GL45 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-247-GL45

missing translation for 'validMainframeMsg'

Designed to fit any reservoir or bottle with GL 45 screw threads.

  • Provides the best possible seal
  • PTFE cap body is chemically inert and remains non-brittle at sub-zero temperatures
  • TFE/propylene o-ring, size 216
  • Polypropylene screw collar
TRUSTED_SUSTAINABILITY

Specifications

Closure Size 45 mm
Liner Material PTFE
Pre-sterilized No
Product Type Screw Cap
Autoclavable Autoclavable
Material PBT
Color Red
No. per Case Cs. of 6
Sterility Non-sterile
For Use With (Equipment) GL 45 Threaded Bottles and Reservoirs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.