Learn More
Fisherbrand™ Glucose Tolerance Test Beverages
GSA_VA
Catalog No. 401009FB Shop All Fisherbrand Products
Change view
Click to view available options
Flavor:
Fruit Punch
Lemon-Lime
Orange
Quantity:
50 g
75 g
100 g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor Quantity
40-100-9FB Lemon-Lime 100 g
40-102-5FB Lemon-Lime 75 g
40-107-4FB Lemon-Lime 50 g
40-120-7FB Orange 100 g
40-127-2FB Orange 50 g
40-150-3FB Fruit Punch 100 g
40-157-6FB Fruit Punch 50 g
40-152-6FB Fruit Punch 75 g
40-122-3FB Orange 75 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 40-100-9FB
This item is not returnable. View return policy
Used to screen for hyperglycemia (diabetes mellitus and gestational diabetes) and hypoglycemia (low blood sugar), these non-carbonated beverages are available in 50, 75 and 100 gram concentrations and orange, lemon-lime and fruit punch flavors.
  • Graduated bottles assure proper dose every time
  • 24-month shelf life
  • Gluten free
Order Info
Packaged in cases of 24 shatter-proof plastic bottles
Certifications
Kosher
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Kosher
Flavor Lemon-Lime
Quantity 100 g