Description
Used to screen for hyperglycemia (diabetes mellitus and gestational diabetes) and hypoglycemia (low blood sugar), these non-carbonated beverages are available in 50, 75 and 100 gram concentrations and orange, lemon-lime and fruit punch flavors.
- Graduated bottles assure proper dose every time
- 24-month shelf life
- Gluten free
Order Info
Packaged in cases of 24 shatter-proof plastic bottles
Certifications
Kosher
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | Kosher |
| Flavor | Fruit Punch |
| Quantity | 50 g |