Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Institutional Trash Can Liners
SDP

Catalog No. 14828400 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
12 to 16 gal.
20 to 30 gal.
32 to 33 gal.
33 gal.
40 to 45 gal.
56 gal.
60 gal.
7 to 10 gal.
Color:
Brown
Clear
Material:
HDPE
LLDPE
Thickness (English):
0.35 mil
0.47 mil
0.63 mil
0.65 mil
0.80 mil
1.15 mil
1.50 mil
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color Material Thickness (English)
14-828-400 7 to 10 gal. Clear LLDPE 0.35 mil
14-828-405 32 to 33 gal. Brown LLDPE 0.65 mil
14-828-404 32 to 33 gal. Clear LLDPE 0.65 mil
14-828-406 32 to 33 gal. Clear LLDPE 1.15 mil
14-828-407 40 to 45 gal. Clear LLDPE 0.80 mil
14-828-408 40 to 45 gal. Clear LLDPE 1.15 mil
14-828-409 60 gal. Clear LLDPE 0.80 mil
14-828-410 56 gal. Clear LLDPE 1.15 mil
14-828-411 56 gal. Brown LLDPE 1.50 mil
14-828-417 33 gal. Clear HDPE 0.63 mil
14-828-419 40 to 45 gal. Clear HDPE 0.63 mil
14-828-401 12 to 16 gal. Clear LLDPE 0.35 mil
14-828-402 20 to 30 gal. Clear LLDPE 0.65 mil
14-828-418 40 to 45 gal. Clear HDPE 0.47 mil
14-828-420 60 gal. Clear HDPE 0.63 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Not for use with biohazardous waste.
Catalog No. 14-828-400
This item is not returnable. View return policy
Not for use with biohazardous waste.

Provide a leak resistant containment solution for non-medical waste disposal

  • For collecting and containing refuse
  • Available in both linear low density (LLDPE) and high density (HDPE) polyethylene film
  • Sold in flat packs for easy dispensing
  • Provide a cost-effective solution for every application

Specifications

Color Clear
For Use With (Application) Collecting and containing refuse, non medical waste disposal
Material LLDPE
Thickness (English) 0.35 mil
Product Type Institutional Trash Can Liner
Capacity (English) 7 to 10 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.