Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lab Automation Pipet Tips
GREENER_CHOICE

Catalog No. 02707262 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
100 μL
1000 μL
20 μL
200 μL
5 to 200 μL
50 μL
Color:
Black
Natural
Sterility:
Non-sterile
Sterile
For Use With (Equipment):
Tecan Genesis
Zymark/Caliper RapidPlate 96, Zymark/Caliper RapidPlate 96/384
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Sterility For Use With (Equipment)
02-707-262 1000 μL Black Sterile Tecan Genesis
02-707-224 100 μL Natural Non-sterile Zymark/Caliper RapidPlate 96, Zymark/Caliper RapidPlate 96/384
02-707-246 20 μL Black Non-sterile Tecan Genesis
02-707-251 50 μL Black Non-sterile Tecan Genesis
02-707-254 200 μL Natural Non-sterile Tecan Genesis
02-707-256 5 to 200 μL Black Non-sterile Tecan Genesis
02-707-259 1000 μL Natural Non-sterile Tecan Genesis
02-707-261 1000 μL Black Non-sterile Tecan Genesis
02-707-219 200 μL Natural Non-sterile Zymark/Caliper RapidPlate 96, Zymark/Caliper RapidPlate 96/384
02-707-220 200 μL Natural Sterile Zymark/Caliper RapidPlate 96, Zymark/Caliper RapidPlate 96/384
02-707-244 20 μL Natural Non-sterile Tecan Genesis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 02-707-262
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provide maximum performance, accuracy and reproducibility in laboratory automation procedures.

Tips are constructed of a highly conductive material necessary to detect the presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.

  • Disposable tips for robotic workstations provide maximum performance, accuracy and reproducibility in laboratory automation procedures
  • Available in clear, blue tint, or carbon-black-conductive polypropylene
  • Carbon-black, liquid-level-sensing tips are required by many automated systems
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Autoclavable Autoclavable
For Use With (Application) Liquid-level-sensing to detect presence or absence of sufficient liquid, provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Format Rack
Tip Style Automation
Disposable Single-use
For Use With (Equipment) Tecan Genesis
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.