Learn More
Description
- Fast drying—highly resistant to abrasion and water
- Designed to write on cold and damp surfaces as easily as they write on dry surfaces
- Will not smear when subjected to water or alcohol during normal use
- Ink is permanent on plastic, wood, and ceramic
- Washes off glass, metal, or porcelain surfaces
Order Info
Sold in packs of 10
Specifications
Specifications
| Product Type | Lab Permanent Marker |
| Tip Style | Fine |
| Color | Blue |
| Sterility | Non-sterile |
| Latex Free | Yes |
| For Use With (Application) | Used to make permanent markings on plastic, metal, or porcelain labware |
| Quantity | 10 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
