Learn More
Bio Plas Microcentrifuge Tube Accessory Micro Tube Tool
Catalog No. 0540722
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
05-407-22 4/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 05-407-22 Supplier Bio Plas Supplier No. 4000
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use in capping or decapping Microcentrifuge Tubes with Snap Caps
  • Polypropylene tool has a slot and tab at one end for use with snap-cap tubes
  • The other end is a socket wrench for screw-cap tubes
Specifications
Material Polypropylene
Quantity 4/Pk.
For Use With (Equipment) One end for snap cap tubes, other end is socket wrench for screw cap tubes, for capping/decapping